Merck
  • A comparative study of the immune modulating properties of antifibrinolytics in cardiac surgery.
A comparative study of the immune modulating properties of antifibrinolytics in cardiac surgery.
Cytokine (2012-11-29)
A F L Later, E F Bruggemans, F P H T M Romijn, J van Pelt, R J M Klautz
ABSTRACT
Antifibrinolytics, used in cardiac surgery to abate postoperative blood loss, share anti-inflammatory properties by suppression of pro-inflammatory D-dimer and plasmin levels. Additional drug specific immune modulating qualities are often mentioned in the discussion on which antifibrinolytic can best be used. To determine the extent and relevance of these effects, we investigated cytokine and growth factor plasma levels in cardiac surgery patients randomized to receive either tranexamic acid, aprotinin, or placebo. Corticosteroid-treated patients served to put the effects in perspective. Using a biochip immunoassay, plasma of 36 cardiac surgery patients was quantified for 12 cytokines and growth factors, assessed preoperatively and 6, 12, 24, and 48 h after the start of cardiopulmonary bypass. Eight patients were treated with tranexamic acid, nine with aprotinin, and nine received placebo. Ten placebo-treated patients received corticosteroids. IL-1ß, IL-6, IL-8, IL-10, IFN-γ, TNF-α, VEGF, MCP-1, and EGF plasma concentrations significantly changed over time across all patients. Aprotinin-treated patients showed decreased pro-inflammatory TNF-α and peak MCP-1 plasma levels when compared with placebo. However, corticosteroids attenuated the inflammatory response to a much larger extent, lowering postoperative IL-6, IL-10, IFN-γ, and VEGF concentrations also. Aprotinin attenuates postoperative pro-inflammatory levels TNF-α and MCP-1 whereas tranexamic acid does not. The majority of plasma proteins studied, however, were not affected by the use of antifibrinolytics when compared with placebo. A clinically relevant common anti-inflammatory effect through inhibition of fibrinolysis seems therefore unlikely.
Sigma-Aldrich
Aprotinin from bovine lung, saline solution, 3-7 TIU/mg protein
Sigma-Aldrich
Aprotinin from bovine lung, lyophilized powder, 3-8 TIU/mg solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Aprotinin from bovine lung, lyophilized powder, 3-7 TIU/mg solid
Sigma-Aldrich
Aprotinin from bovine lung
Sigma-Aldrich
Aprotinin from bovine lung, lyophilized powder, 3-8 TIU/mg solid
Sigma-Aldrich
Aprotinin from bovine lung, BioUltra, 3-8 TIU/mg solid, ≥98% (SDS-PAGE)
Sigma-Aldrich
Aprotinin from bovine lung, lyophilized, ~80% (HPCE), crystalline (fine), white, ≥3500 U/mg