Merck
  • [Simultaneous determination of fumonisins B1 and B2 in traditional Chinese medicines by high-performance liquid chromatography-tandem mass spectrometry].
[Simultaneous determination of fumonisins B1 and B2 in traditional Chinese medicines by high-performance liquid chromatography-tandem mass spectrometry].
Yao xue xue bao = Acta pharmaceutica Sinica (2011-10-21)
Ting-Ting Xie, Feng Qiu, Mei-Hua Yang, Ai-Di Qi
ABSTRACT
A rapid and sensitive analytical method was developed for the simultaneous determination of fumonisins B1 and B2 in traditional Chinese medicines by HPLC-MS/MS. The detection limits for fumonisins B1 and B2 were 0.25 ng x mL(-1) corresponding to 2 microg x kg(-1) in samples. Recoveries from different samples spiked with fumonisins B1 and B2 at levels ranging from 0.2 to 3 mg x kg(-1) were 84.0%-96.1% and 86.3%-99.3%, respectively. Among the total of 34 samples purchased from local markets, ten samples of which were visibly moldy samples due to inappropriate storage, and 24 were normal samples. The results showed that 6 of the visibly moldy samples and 5 of the normal samples were contaminated with total fumonisins at levels ranging 82.4-2349 microg x kg(-1) and 102-729 microg x kg(-1), respectively.
Sigma-Aldrich
Fumonisin B2 from Fusarium moniliforme, sphingosine N-acyltransferase inhibitor
SKU
Pack Size
Availability
Price
Quantity
Supelco
Fumonisin B2 solution, ~50 μg/mL in acetonitrile: water (50:50), analytical standard