Merck
  • Correlation of toxicity with lead content in root tip cells (Allium cepa L.).
Correlation of toxicity with lead content in root tip cells (Allium cepa L.).
Biological trace element research (2008-07-19)
Ingrid Carruyo, Yusmary Fernández, Letty Marcano, Xiomara Montiel, Zaida Torrealba
ABSTRACT
The present study determines lead content in onion root tip cells (Allium cepa L.), correlating it with its toxicity. The treatment was carried at 25 +/- 0.5 degrees C using aqueous solutions of lead chloride at 0.1, 0.25, 0.50, 0.75, and 1 ppm for 12, 24, 48, and 72 h. For each treatment, a control where the lead solution was substituted by distilled water was included. After treatment, the meristems were fixed with a mixture of alcohol-acetic acid (3:1) and colored according to the technique of Feulgen. Lead content was quantified by graphite furnace absorption atomic spectrometry. The lead content in the roots ranged from 3.25 to 244.72 microg/g dry weight, with a direct relation with the concentration and time of exposure. A significant negative correlation was presented (r = -0.3629; p < 0.01) among lead content and root growth increment, and a positive correlation (r = 0.7750; p < 0.01) with the induction of chromosomic aberrations. In conclusion, lead is able to induce a toxic effect in the exposed roots, correlated with its content.
Sigma-Aldrich
Lead(II) chloride, AnhydroBeads, −10 mesh, 99.999%
Sigma-Aldrich
Lead(II) chloride, 99.999% trace metals basis
Sigma-Aldrich
Lead(II) chloride, powder, 98%