Merck
  • Synthesis of carbohydrates in a continuous flow reactor by immobilized phosphatase and aldolase.
Synthesis of carbohydrates in a continuous flow reactor by immobilized phosphatase and aldolase.
ChemSusChem (2012-11-15)
Lara Babich, Aloysius F Hartog, Lieke J C van Hemert, Floris P J T Rutjes, Ron Wever
ABSTRACT
Herein, we report a new flow process with immobilized enzymes to synthesize complex chiral carbohydrate analogues from achiral inexpensive building blocks in a three-step cascade reaction. The first reactor contained immobilized acid phosphatase, which phosphorylated dihydroxyacetone to dihydroxyacetone phosphate using pyrophosphate as the phosphate donor. The second flow reactor contained fructose-1,6-diphosphate aldolase (RAMA, rabbit muscle aldolase) or rhamnulose-1-phosphate aldolase (RhuA from Thermotoga maritima) and acid phosphatase. The immobilized aldolases coupled the formed dihydroxyacetone phosphate to aldehydes, resulting in phosphorylated carbohydrates. A final reactor containing acid phosphatase that dephosphorylated the phosphorylated product yielded the final product. Different aldehydes were used to synthesize carbohydrates on a gram scale. To demonstrate the feasibility of the flow systems, we synthesized 0.6 g of the D-fagomine precursor. By using immobilized aldolase RhuA we were also able to obtain other stereoisomers of the D-fagomine precursor.
Sigma-Aldrich
Dihydroxyacetone phosphate hemimagnesium salt hydrate, ≥95% (TLC)
Sigma-Aldrich
Aldolase from rabbit muscle, ammonium sulfate suspension, 10-20 units/mg protein
Sigma-Aldrich
Aldolase from rabbit muscle, lyophilized powder, ≥8.0 units/mg protein