Merck
  • Increase in Tau Pathology in P290S Mapt Knock-In Mice Crossed with App NL-G-F Mice.
Increase in Tau Pathology in P290S Mapt Knock-In Mice Crossed with App NL-G-F Mice.
eNeuro (2023-01-13)
Melissa Huang, Jennifer Macdonald, Isabelle Lavenir, Renren Chen, Molly Craxton, Elizabeth Slavik-Smith, Stephen W Davies, Michel Goedert
ABSTRACT
Alzheimer's Disease (AD) is characterized by the pathologic assembly of amyloid β (Aβ) peptide, which deposits into extracellular plaques, and tau, which accumulates in intraneuronal inclusions. To investigate the link between Aβ and tau pathologies, experimental models featuring both pathologies are needed. We developed a mouse model featuring both tau and Aβ pathologies by knocking the P290S mutation into murine Mapt and crossing these Mapt P290S knock-in (KI) mice with the App NL-G-F KI line. Mapt P290S KI mice developed a small number of tau inclusions, which increased with age. The amount of tau pathology was significantly larger in App NL-G-F xMapt P290S KI mice from 18 months of age onward. Tau pathology was higher in limbic areas, including hippocampus, amygdala, and piriform/entorhinal cortex. We also observed AT100-positive and Gallyas-Braak-silver-positive dystrophic neurites containing assembled filamentous tau, as visualized by in situ electron microscopy. Using a cell-based tau seeding assay, we showed that Sarkosyl-insoluble brain extracts from both 18-month-old Mapt P290S KI and App NL-G-F xMapt P290S KI mice were seed competent, with brain extracts from double-KI mice seeding significantly more than those from the Mapt P290S KI mice. Finally, we showed that App NL-G-F xMapt P290S KI mice had neurodegeneration in the piriform cortex from 18 months of age. We suggest that App NL-G-F xMapt P290S KI mice provide a good model for studying the interactions of aggregation-prone tau, Aβ, neuritic plaques, neurodegeneration, and aging.
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-APP A4 Antibody, a.a. 66-81 of APP {NT}, clone 22C11, clone 22C11, Chemicon®, from mouse
Sigma-Aldrich
Anti-Tau Antibody, clone T49 (Not human), clone T49, from mouse
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Gold antibody produced in goat, affinity isolated antibody, aqueous glycerol suspension, 10 nm (colloidal gold)