Merck
  • Brain-derived neurotrophic factor stimulates proliferation and differentiation of neural stem cells, possibly by triggering the Wnt/β-catenin signaling pathway.
Brain-derived neurotrophic factor stimulates proliferation and differentiation of neural stem cells, possibly by triggering the Wnt/β-catenin signaling pathway.
Journal of neuroscience research (2012-10-02)
Bei-Yu Chen, Xi Wang, Zhao-Yan Wang, Ya-Zhou Wang, Liang-Wei Chen, Zhuo-Jing Luo
ABSTRACT
Brain-derived neurotrophic factor (BDNF) has critical functions in promoting survival, expansion, and differentiation of neural stem cells (NSCs), but its downstream regulation mechanism is still not fully understood. The role of BDNF in proliferation and differentiation of NSCs through Wnt/β-catenin signaling was studied via cell culture of cortical NSCs, Western blotting, immunocytochemistry, and TOPgal (Wnt reporter) analysis in mice. First, BDNF stimulated NSC proliferation dose dependently in cultured neurospheres that exhibited BrdU incorporation and neuronal and glial differentiation abilities. Second, BDNF effectively enhanced cell commitment to neuronal and oligodendrocytic fates, as indicated by increased differentiation marker Tuj-1 (neuronal marker), CNPase (oligodendrocyte marker), and neuronal process extension. Third, BDNF upregulated expression of Wnt/β-catenin signaling (Wnt1 and free β-catenin) molecules. Moreover, these promoting effects were significantly inhibited by application of IWR1, a Wnt signaling-specific blocker in culture. The TOPgal mouse experiment further confirmed BDNF-triggered Wnt signaling activation by β-gal labeling. Finally, an MEK inhibition experiment showed a mediating role of the microtubule-associated protein kinase pathway in BDNF-triggered Wnt/β-catenin signaling cascades. This study overall has revealed that BDNF might contribute to proliferation and neuronal and oligodendrocytic differentiation of NSCs in vitro, most possibly by triggering the Wnt/β-catenin signaling pathway. Nevertheless, determining the exact cross-talk points at which BDNF might stimulate Wnt/β-catenin signaling pathway in NSC activity requires further investigation.
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, ascites fluid, clone GA5, Chemicon®
Sigma-Aldrich
Anti-β Galactosidase Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-BrdU Antibody, clone PRB-1, FITC conjugated, clone PRB-1, Chemicon®, from mouse
Sigma-Aldrich
Anti-β-Tubulin III antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
PD 98,059, solid