Merck
  • Mechanism of integrin activation by talin and its cooperation with kindlin.
Mechanism of integrin activation by talin and its cooperation with kindlin.
Nature communications (2022-04-30)
Fan Lu, Liang Zhu, Thomas Bromberger, Jun Yang, Qiannan Yang, Jianmin Liu, Edward F Plow, Markus Moser, Jun Qin
ABSTRACT
Talin-induced integrin binding to extracellular matrix ligands (integrin activation) is the key step to trigger many fundamental cellular processes including cell adhesion, cell migration, and spreading. Talin is widely known to use its N-terminal head domain (talin-H) to bind and activate integrin, but how talin-H operates in the context of full-length talin and its surrounding remains unknown. Here we show that while being capable of inducing integrin activation, talin-H alone exhibits unexpectedly low potency versus a constitutively activated full-length talin. We find that the large C-terminal rod domain of talin (talin-R), which otherwise masks the integrin binding site on talin-H in inactive talin, dramatically enhances the talin-H potency by dimerizing activated talin and bridging it to the integrin co-activator kindlin-2 via the adaptor protein paxillin. These data provide crucial insight into the mechanism of talin and its cooperation with kindlin to promote potent integrin activation, cell adhesion, and signaling.
Sigma-Aldrich
Anti-Actin (Ab-1) Mouse mAb (JLA20), liquid, clone JLA20, Calbiochem®
Sigma-Aldrich
Anti-Kindlin-2 Antibody, clone 3A3, clone 3A3, from mouse