Merck
  • Chromatin immunoprecipitation (ChIP) coupled to detection by quantitative real-time PCR to study transcription factor binding to DNA in Caenorhabditis elegans.
Chromatin immunoprecipitation (ChIP) coupled to detection by quantitative real-time PCR to study transcription factor binding to DNA in Caenorhabditis elegans.
Nature protocols (2008-04-05)
Arnab Mukhopadhyay, Bart Deplancke, Albertha J M Walhout, Heidi A Tissenbaum
ABSTRACT
In order to determine how signaling pathways differentially regulate gene expression, it is necessary to identify the interactions between transcription factors (TFs) and their cognate cis-regulatory DNA elements. Here, we have outlined a chromatin immunoprecipitation (ChIP) protocol for use in whole Caenorhabditis elegans extracts. We discuss optimization of the procedure, including growth and harvesting of the worms, formaldehyde fixation, TF immunoprecipitation and analysis of bound sequences through real-time PCR. It takes approximately 10-12 d to obtain the worm culture for ChIP; the ChIP procedure is spaced out over a period of 2.5 d with two overnight incubations.
Product Description
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Millipore
NP-40 Alternative
Sigma-Aldrich
Protease Inhibitor Cocktail powder, for general use, lyophilized powder