Merck
  • G3BPs tether the TSC complex to lysosomes and suppress mTORC1 signaling.
G3BPs tether the TSC complex to lysosomes and suppress mTORC1 signaling.
Cell (2021-01-27)
Mirja Tamara Prentzell, Ulrike Rehbein, Marti Cadena Sandoval, Ann-Sofie De Meulemeester, Ralf Baumeister, Laura Brohée, Bianca Berdel, Mathias Bockwoldt, Bernadette Carroll, Suvagata Roy Chowdhury, Andreas von Deimling, Constantinos Demetriades, Gianluca Figlia, Mariana Eca Guimaraes de Araujo, Alexander M Heberle, Ines Heiland, Birgit Holzwarth, Lukas A Huber, Jacek Jaworski, Magdalena Kedra, Katharina Kern, Andrii Kopach, Viktor I Korolchuk, Ineke van 't Land-Kuper, Matylda Macias, Mark Nellist, Wilhelm Palm, Stefan Pusch, Jose Miguel Ramos Pittol, Michèle Reil, Anja Reintjes, Friederike Reuter, Julian R Sampson, Chloë Scheldeman, Aleksandra Siekierska, Eduard Stefan, Aurelio A Teleman, Laura E Thomas, Omar Torres-Quesada, Saskia Trump, Hannah D West, Peter de Witte, Sandra Woltering, Teodor E Yordanov, Justyna Zmorzynska, Christiane A Opitz, Kathrin Thedieck
ABSTRACT
Ras GTPase-activating protein-binding proteins 1 and 2 (G3BP1 and G3BP2, respectively) are widely recognized as core components of stress granules (SGs). We report that G3BPs reside at the cytoplasmic surface of lysosomes. They act in a non-redundant manner to anchor the tuberous sclerosis complex (TSC) protein complex to lysosomes and suppress activation of the metabolic master regulator mechanistic target of rapamycin complex 1 (mTORC1) by amino acids and insulin. Like the TSC complex, G3BP1 deficiency elicits phenotypes related to mTORC1 hyperactivity. In the context of tumors, low G3BP1 levels enhance mTORC1-driven breast cancer cell motility and correlate with adverse outcomes in patients. Furthermore, G3bp1 inhibition in zebrafish disturbs neuronal development and function, leading to white matter heterotopia and neuronal hyperactivity. Thus, G3BPs are not only core components of SGs but also a key element of lysosomal TSC-mTORC1 signaling.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
RIPA Buffer
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sigma-Aldrich
Doxycycline hydrochloride
Sigma-Aldrich
Rapamycin, ≥95% (HPLC), powder
Sigma-Aldrich
4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride, ≥97.0% (HPLC)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
IgG from rabbit serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder
Sigma-Aldrich
E-64, protease inhibitor
Sigma-Aldrich
Benzamidine hydrochloride hydrate, ≥99%
Sigma-Aldrich
Magnesium chloride hexahydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Fetal Bovine Serum, Heat Inactivated, non-USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
AptaFluor® SAH Methyltransferase Assay
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in a glass vial
Sigma-Aldrich
Triton X-100 solution, BioUltra, ~10% in H2O
Sigma-Aldrich
1,4-Diazabicyclo[2.2.2]octane, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethyl 3-aminobenzoate methanesulfonate, 98%
Sigma-Aldrich
IGEPAL® CA-630
Sigma-Aldrich
Insulin from bovine pancreas, γ-irradiated, BioXtra, suitable for cell culture, potency: ≥20 units/mg (USP units), lyophilized powder
Sigma-Aldrich
Potassium chloride, ≥99.0%
Sigma-Aldrich
Aprotinin from bovine lung, lyophilized powder, 3-8 TIU/mg solid
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Hexadimethrine bromide, ≥94% (titration)
Sigma-Aldrich
Bromophenol Blue sodium salt
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)