Merck
  • Steroid Receptor Coactivator 3 Regulates Synaptic Plasticity and Hippocampus-dependent Memory.
Steroid Receptor Coactivator 3 Regulates Synaptic Plasticity and Hippocampus-dependent Memory.
Neuroscience bulletin (2021-07-07)
Hai-Long Zhang, Bing Zhao, Pin Yang, Yin-Quan Du, Wei Han, Jianming Xu, Dong-Min Yin
ABSTRACT
Steroid hormones play important roles in brain development and function. The signaling of steroid hormones depends on the interaction between steroid receptors and their coactivators. Although the function of steroid receptor coactivators has been extensively studied in other tissues, their functions in the central nervous system are less well investigated. In this study, we addressed the function of steroid receptor coactivator 3 (SRC3) - a member of the p160 SRC protein family that is expressed predominantly in the hippocampus. While hippocampal development was not altered in Src3+/- mice, hippocampus-dependent functions such as short-term memory and spatial memory were impaired. We further demonstrated that the deficient learning and memory in Src3+/- mice was strongly associated with the impairment of long-term potentiation (LTP) at Schaffer Collateral-CA1 synapses. Mechanistic studies indicated that Src3+/- mutation altered the composition of N-methyl-D-aspartate receptor subunits in the postsynaptic densities of hippocampal neurons. Finally, we showed that SRC3 regulated synaptic plasticity and learning mainly dependent on its lysine acetyltransferase activity. Taken together, these results reveal previously unknown functions of SRC3 in the hippocampus and thus may provide insight into how steroid hormones regulate brain function.
Sigma-Aldrich
Anti-Post Synaptic Density Protein 95 Antibody, clone 6G6-1C9, clone 6G6-1C9, Chemicon®, from mouse
Sigma-Aldrich
Anti-GluR2 Antibody, clone L21/32, clone L21/32, from mouse
Sigma-Aldrich
Anti-NMDAR2B Antibody, CT, ascites fluid, clone 1C6.5C4, Chemicon®
Sigma-Aldrich
Anti-Calmodulin Antibody, Upstate®, from mouse
Sigma-Aldrich
Anti-NR2A Antibody, clone A12W, rabbit monoclonal, culture supernatant, clone A12W, Upstate®
Sigma-Aldrich
Biocytin, ≥98% (TLC)