Merck
  • Regional identity of human neural stem cells determines oncogenic responses to histone H3.3 mutants.
Regional identity of human neural stem cells determines oncogenic responses to histone H3.3 mutants.
Cell stem cell (2021-02-26)
Raul Bardini Bressan, Benjamin Southgate, Kirsty M Ferguson, Carla Blin, Vivien Grant, Neza Alfazema, Jimi C Wills, Maria Angeles Marques-Torrejon, Gillian M Morrison, James Ashmore, Faye Robertson, Charles A C Williams, Leanne Bradley, Alex von Kriegsheim, Richard A Anderson, Simon R Tomlinson, Steven M Pollard
ABSTRACT
Point mutations within the histone H3.3 are frequent in aggressive childhood brain tumors known as pediatric high-grade gliomas (pHGGs). Intriguingly, distinct mutations arise in discrete anatomical regions: H3.3-G34R within the forebrain and H3.3-K27M preferentially within the hindbrain. The reasons for this contrasting etiology are unknown. By engineering human fetal neural stem cell cultures from distinct brain regions, we demonstrate here that cell-intrinsic regional identity provides differential responsiveness to each mutant that mirrors the origins of pHGGs. Focusing on H3.3-G34R, we find that the oncohistone supports proliferation of forebrain cells while inducing a cytostatic response in the hindbrain. Mechanistically, H3.3-G34R does not impose widespread transcriptional or epigenetic changes but instead impairs recruitment of ZMYND11, a transcriptional repressor of highly expressed genes. We therefore propose that H3.3-G34R promotes tumorigenesis by focally stabilizing the expression of key progenitor genes, thereby locking initiating forebrain cells into their pre-existing immature state.
Sigma-Aldrich
Senescence Cells Histochemical Staining Kit, sufficient for 100 tests
Sigma-Aldrich
Anti-HIRA Antibody, clone WC119, clone WC119, from mouse
Millipore
Benzonase® Nuclease, ≥250 units/μL, ≥90% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in a glass vial
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys4) Antibody, Upstate®, from rabbit
Millipore
FluorSave Reagent
Sigma-Aldrich
Anti-Histone H3.3 Antibody, from rabbit, purified by affinity chromatography