Merck
  • Tau activates microglia via the PQBP1-cGAS-STING pathway to promote brain inflammation.
Tau activates microglia via the PQBP1-cGAS-STING pathway to promote brain inflammation.
Nature communications (2021-11-17)
Meihua Jin, Hiroki Shiwaku, Hikari Tanaka, Takayuki Obita, Sakurako Ohuchi, Yuki Yoshioka, Xiaocen Jin, Kanoh Kondo, Kyota Fujita, Hidenori Homma, Kazuyuki Nakajima, Mineyuki Mizuguchi, Hitoshi Okazawa
ABSTRACT
Brain inflammation generally accompanies and accelerates neurodegeneration. Here we report a microglial mechanism in which polyglutamine binding protein 1 (PQBP1) senses extrinsic tau 3R/4R proteins by direct interaction and triggers an innate immune response by activating a cyclic GMP-AMP synthase (cGAS)-Stimulator of interferon genes (STING) pathway. Tamoxifen-inducible and microglia-specific depletion of PQBP1 in primary culture in vitro and mouse brain in vivo shows that PQBP1 is essential for sensing-tau to induce nuclear translocation of nuclear factor κB (NFκB), NFκB-dependent transcription of inflammation genes, brain inflammation in vivo, and eventually mouse cognitive impairment. Collectively, PQBP1 is an intracellular receptor in the cGAS-STING pathway not only for cDNA of human immunodeficiency virus (HIV) but also for the transmissible neurodegenerative disease protein tau. This study characterises a mechanism of brain inflammation that is common to virus infection and neurodegenerative disorders.
Product Description
Millipore
Protease Inhibitor Cocktail Set III, EDTA-Free, Protease inhibitor cocktail III, EDTA-free for inhibiting aspartic, cysteine, and serine proteases as well as aminopeptidases in mammalian cells and tissues.
Sigma-Aldrich
Anti-Tau Antibody, a.a. 210-241, clone Tau-5, ascites fluid, clone Tau-5, Chemicon®
Sigma-Aldrich
1-Butyl-3-methylimidazolium hexafluorophosphate, ≥97.0% (HPLC)
Sigma-Aldrich
4-Hydroxytamoxifen Ready Made Solution, 5 mg/mL in ethanol: isopropanol (95:5)
Sigma-Aldrich
Anti-cGAS Antibody, from rabbit, purified by affinity chromatography
Roche
Biotin-16-dUTP
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Corn oil, delivery vehicle for fat-soluble compounds