Merck
  • Human skeletal muscle CD90+ fibro-adipogenic progenitors are associated with muscle degeneration in type 2 diabetic patients.
Human skeletal muscle CD90+ fibro-adipogenic progenitors are associated with muscle degeneration in type 2 diabetic patients.
Cell metabolism (2021-10-23)
Jean Farup, Jesper Just, Frank de Paoli, Lin Lin, Jonas Brorson Jensen, Tine Billeskov, Ines Sanchez Roman, Cagla Cömert, Andreas Buch Møller, Luca Madaro, Elena Groppa, Rikard Göran Fred, Ulla Kampmann, Lars C Gormsen, Steen B Pedersen, Peter Bross, Tinna Stevnsner, Nikolaj Eldrup, Tune H Pers, Fabio M V Rossi, Pier Lorenzo Puri, Niels Jessen
ABSTRACT
Type 2 diabetes mellitus (T2DM) is associated with impaired skeletal muscle function and degeneration of the skeletal muscles. However, the mechanisms underlying the degeneration are not well described in human skeletal muscle. Here we show that skeletal muscle of T2DM patients exhibit degenerative remodeling of the extracellular matrix that is associated with a selective increase of a subpopulation of fibro-adipogenic progenitors (FAPs) marked by expression of THY1 (CD90)-the FAPCD90+. We identify platelet-derived growth factor (PDGF) as a key FAP regulator, as it promotes proliferation and collagen production at the expense of adipogenesis. FAPsCD90+ display a PDGF-mimetic phenotype, with high proliferative activity, clonogenicity, and production of extracellular matrix. FAPCD90+ proliferation was reduced by in vitro treatment with metformin. Furthermore, metformin treatment reduced FAP content in T2DM patients. These data identify a PDGF-driven conversion of a subpopulation of FAPs as a key event in the fibrosis development in T2DM muscle.
Sigma-Aldrich
Anti-Fibroblasts Antibody, clone TE-7, clone TE-7, Chemicon®, from mouse
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), crystalline
Sigma-Aldrich
hBFGF, FGF-Basic, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Transforming Growth Factor-β1 human, TGF-β1, recombinant, expressed in CHO cells, powder, suitable for cell culture
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, BioUltra, ≥99%
Sigma-Aldrich
ECM Gel from Engelbreth-Holm-Swarm murine sarcoma, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Nutrient Mixture F-10 Ham, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
1,1-Dimethylbiguanide hydrochloride, 97%
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)
Sigma-Aldrich
D-(+)-Glucose solution, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
SAFC
Insulin, Human Recombinant, for research or for further manufacturing use, dry powder
Sigma-Aldrich
Imatinib mesylate, ≥98% (HPLC)
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
Alizarin Red S, certified by the BSC
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture