Merck
  • Genome-wide Fitness Profiles Reveal a Requirement for Autophagy During Yeast Fermentation.
Genome-wide Fitness Profiles Reveal a Requirement for Autophagy During Yeast Fermentation.
G3 (Bethesda, Md.) (2012-03-03)
Nina Piggott, Michael A Cook, Mike Tyers, Vivien Measday
ABSTRACT
The ability of cells to respond to environmental changes and adapt their metabolism enables cell survival under stressful conditions. The budding yeast Saccharomyces cerevisiae (S. cerevisiae) is particularly well adapted to the harsh conditions of anaerobic wine fermentation. However, S. cerevisiae gene function has not been previously systematically interrogated under conditions of industrial fermentation. We performed a genome-wide study of essential and nonessential S. cerevisiae gene requirements during grape juice fermentation to identify deletion strains that are either depleted or enriched within the viable fermentative population. Genes that function in autophagy and ubiquitin-proteasome degradation are required for optimal survival during fermentation, whereas genes that function in ribosome assembly and peroxisome biogenesis impair fitness during fermentation. We also uncover fermentation phenotypes for 139 uncharacterized genes with no previously known cellular function. We demonstrate that autophagy is induced early in wine fermentation in a nitrogen-replete environment, suggesting that autophagy may be triggered by other forms of stress that arise during fermentation. These results provide insights into the complex fermentation process and suggest possible means for improvement of industrial fermentation strains.
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
L-(+)-Tartaric acid, BioXtra
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-(−)-Malic acid, ≥95% (titration)