Merck
  • YAP and TAZ regulate Schwann cell proliferation and differentiation during peripheral nerve regeneration.
YAP and TAZ regulate Schwann cell proliferation and differentiation during peripheral nerve regeneration.
Glia (2020-12-19)
Haley Jeanette, Leandro N Marziali, Urja Bhatia, Abigail Hellman, Jacob Herron, Ashley M Kopec, Maria Laura Feltri, Yannick Poitelon, Sophie Belin
ABSTRACT
YAP and TAZ are effectors of the Hippo pathway that controls multicellular development by integrating chemical and mechanical signals. Peripheral nervous system development depends on the Hippo pathway. We previously showed that loss of YAP and TAZ impairs the development of peripheral nerve as well as Schwann cell myelination. The role of the Hippo pathway in peripheral nerve regeneration has just started to be explored. After injury, Schwann cells adopt new identities to promote regeneration by converting to a repair-promoting phenotype. While the reprogramming of Schwann cells to repair cells has been well characterized, the maintenance of such repair phenotype cannot be sustained for a very long period, which limits nerve repair in human. First, we show that short or long-term myelin maintenance is not affected by defect in YAP and TAZ expression. Using crush nerve injury and conditional mutagenesis in mice, we also show that YAP and TAZ are regulators of repair Schwann cell proliferation and differentiation. We found that YAP and TAZ are required in repair Schwann cells for their redifferentiation into myelinating Schwann cell following crush injury. In this present study, we describe how the Hippo pathway and YAP and TAZ regulate remyelination over time during peripheral nerve regeneration.
Sigma-Aldrich
Anti-phospho Histone H3 (Ser10) Antibody, Alexa Fluor 488 Conjugate, from rabbit, ALEXA FLUOR 488
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
DAPI, for nucleic acid staining