Merck
  • STIM1 long and STIM1 gate differently TRPC1 during store-operated calcium entry.
STIM1 long and STIM1 gate differently TRPC1 during store-operated calcium entry.
Cell calcium (2019-12-16)
Agnieszka Dyrda, Stéphane Koenig, Maud Frieden
ABSTRACT
During myogenesis, a long splice variant of STIM1, called STIM1L is getting expressed, while the level of STIM1 remains constant. Previous work demonstrated that STIM1L is more efficient in eliciting store-operated Ca2+ entry (SOCE), but no current analysis of the channel(s) activated by this new STIM1L isoform was performed until now. In this study, we investigate the ionic channel(s) activated by STIM1L and whether differences exist between the two STIM1 isoforms, using HEK-293 T cells as a model system. Our data show that STIM1 and STIM1L activate Orai1 channel but also the endogenously expressed TRPC1. The channel activation occurs in two steps, with first Orai1 activation followed, in a subset of cells, by TRPC1 opening. Remarkably, STIM1L more frequently activates TRPC1 and preferentially interacts with TRPC1. In low intracellular Ca2+ buffering condition, the frequency of TRPC1 opening increases significantly, strongly suggesting a Ca2+-dependent channel activation. The ability of STIM1L to open Orai1 appears decreased compared to STIM1, which might be explained by its stronger propensity towards TRPC1. Indeed, increasing the amount of STIM1L results in an enhanced Orai1 current. The role of endogenous TRPC1 in STIM1- and STIM1L-induced SOCE was confirmed by Ca2+ imaging experiments. Overall, our findings provide a detailed analysis of the channels activated by both STIM1 isoforms, revealing that STIM1L is more prone to open TRPC1, which might explain the larger SOCE elicited by this isoform.
Sigma-Aldrich
Ponceau S solution, for protein sequence analysis, for protein analysis, BioReagent, suitable (for use in cellulose acetate electrophoresis), 0.1 % (w/v) in 5% acetic acid
Sigma-Aldrich
Anti-STIM1 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Fetuin from fetal bovine serum, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film