Merck
  • Fermentation Products of Commensal Bacteria Alter Enterocyte Lipid Metabolism.
Fermentation Products of Commensal Bacteria Alter Enterocyte Lipid Metabolism.
Cell host & microbe (2020-02-27)
João R Araújo, Asmaa Tazi, Odile Burlen-Defranoux, Sophie Vichier-Guerre, Giulia Nigro, Hélène Licandro, Sylvie Demignot, Philippe J Sansonetti
ABSTRACT
Despite the recognized capacity of the gut microbiota to regulate intestinal lipid metabolism, the role of specific commensal species remains undefined. Here, we aimed to understand the bacterial effectors and molecular mechanisms by which Lactobacillus paracasei and Escherichia coli regulate lipid metabolism in enterocytes. We show that L-lactate produced by L. paracasei inhibits chylomicron secretion from enterocytes and promotes lipid storage by a mechanism involving L-lactate absorption by enterocytes, its conversion to malonyl-CoA, and the subsequent inhibition of lipid beta-oxidation. In contrast, acetate produced by E. coli also inhibits chylomicron secretion by enterocytes but promotes lipid oxidation by a mechanism involving acetate absorption by enterocytes, its metabolism to acetyl-CoA and AMP, and the subsequent upregulation of the AMPK/PGC-1α/PPARα pathway. Our study opens perspectives for developing specific bacteria- and metabolite-based therapeutic interventions against obesity, atherosclerosis, and malnutrition by targeting lipid metabolism in enterocytes.
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Anti-FFAR2 Antibody/GPR43, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Glucose (GO) Assay Kit, sufficient for 20 assays
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, ≥98% (TLC), powder
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution