Merck
  • Comparison of AAV-Mediated Optogenetic Vision Restoration between Retinal Ganglion Cell Expression and ON Bipolar Cell Targeting.
Comparison of AAV-Mediated Optogenetic Vision Restoration between Retinal Ganglion Cell Expression and ON Bipolar Cell Targeting.
Molecular therapy. Methods & clinical development (2020-06-18)
Qi Lu, Tushar H Ganjawala, Andrea Krstevski, Gary W Abrams, Zhuo-Hua Pan
ABSTRACT
The loss of photoreceptors in individuals with retinal degenerative diseases leads to partial or complete blindness. Optogenetic therapy is a promising approach for restoring vision to the blind. Multiple strategies have been employed by targeting genetically encoded light sensors, particularly channelrhodopsins, to surviving retinal neurons in animal models. In particular, the strategy of targeting retinal bipolar cells has commonly been expected to result in better vision than ubiquitous expression in retinal ganglion cells. However, a direct comparison of the channelrhodopsin-restored vision between these two strategies has not been performed. Here, we compared the restored visual functions achieved by adeno-associated virus (AAV)-mediated expression of a channelrhodopsin in ON-type bipolar cells and retinal ganglion cells driven by an improved mGluR6 promoter and a CAG promoter, respectively, in a blind mouse model by performing electrophysiological recordings and behavioral assessments. Unexpectedly, the efficacy of the restored vision based on light sensitivity and visual acuity was much higher following ubiquitous retinal ganglion cell expression than that of the strategy targeting ON-type bipolar cells. Our study suggests that, at least based on currently available gene delivery techniques, the expression of genetically encoded light sensors in retinal ganglion cells is likely a practical and advantageous strategy for optogenetic vision restoration.
Sigma-Aldrich
Sample Buffer, Laemmli 2× Concentrate
Sigma-Aldrich
Anti-RBPMS Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution