Merck
  • Ubiquitin-regulated recruitment of IkappaB kinase epsilon to the MAVS interferon signaling adapter.
Ubiquitin-regulated recruitment of IkappaB kinase epsilon to the MAVS interferon signaling adapter.
Molecular and cellular biology (2009-04-22)
Suzanne Paz, Myriam Vilasco, Meztli Arguello, Qiang Sun, Judith Lacoste, Thi Lien-Anh Nguyen, Tiejun Zhao, Elena A Shestakova, Scott Zaari, Annie Bibeau-Poirier, Marc J Servant, Rongtuan Lin, Eliane F Meurs, John Hiscott
ABSTRACT
Induction of the antiviral interferon response is initiated upon recognition of viral RNA structures by the RIG-I or Mda-5 DEX(D/H) helicases. A complex signaling cascade then converges at the mitochondrial adapter MAVS, culminating in the activation of the IRF and NF-kappaB transcription factors and the induction of interferon gene expression. We have previously shown that MAVS recruits IkappaB kinase epsilon (IKKepsilon) but not TBK-1 to the mitochondria following viral infection. Here we map the interaction of MAVS and IKKepsilon to the C-terminal region of MAVS and demonstrate that this interaction is ubiquitin dependent. MAVS is ubiquitinated following Sendai virus infection, and K63-linked ubiquitination of lysine 500 (K500) of MAVS mediates recruitment of IKKepsilon to the mitochondria. Real-time PCR analysis reveals that a K500R mutant of MAVS increases the mRNA level of several interferon-stimulated genes and correlates with increased NF-kappaB activation. Thus, recruitment of IKKepsilon to the mitochondria upon MAVS K500 ubiquitination plays a modulatory role in the cascade leading to NF-kappaB activation and expression of inflammatory and antiviral genes. These results provide further support for the differential role of IKKepsilon and TBK-1 in the RIG-I/Mda5 pathway.
Sigma-Aldrich
Deoxynucleotide Mix, 10 mM, Molecular Biology Reagent
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Deoxynucleotide Set, 10 mM, Individual dNTPs for routine PCR; 0.5 mL each