MilliporeSigma
  • A cell sorter with modified bamboo charcoal for the efficient selection of specific antibody-producing hybridomas.
A cell sorter with modified bamboo charcoal for the efficient selection of specific antibody-producing hybridomas.
Biomaterials (2010-08-17)
Chien-Chen Lin, Mei-Hui Ni, Yu-Chung Chang, Hsiu-Lun Yeh, Feng-Huei Lin
ABSTRACT
Monoclonal antibodies (mAbs) have been proven useful in research and clinical applications. However, the generation of mAbs by conventional hybridoma technology is time-, cost- and labor-consuming. Here we developed a simplified procedure for efficient generation and selection of antibody-producing hybridomas within 1 h, using a particular cell sorter design, a cytoflow reactor-based cell sorter (CBCS) which consists mainly of the "cytoflow reactor" that comprises two components, a reaction chamber and a glass tubing for air and medium exchange by gravity, and the "sorting material", human EGFR-conjugated bamboo charcoal, for specific B-cell enrichment. The high surface area and porous structure of bamboo charcoal greatly increased cell density and protein production. Moreover, from Raman, FT-IR spectroscopy and IFA analysis, the carboxylation and immobilization of bamboo charcoal can be introduced easily by nitric acid treatment and conjugated handily with human EGFR using EDC/NHS. Other evidences, such as IFA, showed that the specific hybridomas generated in this study could secrete specific anti-human EGFR antibodies. Our design allows the production of mAbs while avoiding time-consuming steps, such as large numbers of limiting dilutions and screening assays, and demonstrates that the CBCS could be a powerful tool for monoclonal antibody production.
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, BioXtra