Merck
  • Reversion-inducing cysteine-rich protein with Kazal motifs interferes with epidermal growth factor receptor signaling.
Reversion-inducing cysteine-rich protein with Kazal motifs interferes with epidermal growth factor receptor signaling.
Oncogene (2010-10-05)
S Kitajima, T Miki, Y Takegami, Y Kido, M Noda, E Hara, A Shamma, C Takahashi
ABSTRACT
The reversion-inducing cysteine-rich protein with Kazal motifs (RECK) gene had been isolated as an antagonist to RAS signaling; however, the mechanism of its action is not clear. In this study, the effect of loss of RECK function was assessed in various ways and cell systems. Successive cell cultivation of mouse embryonic fibroblasts (MEFs) according to 3T3 protocol revealed that the germline knockout of RECK confers accelerated cell proliferation and early escape from cellular senescence associated with downregulation of p19(Arf), Trp53 and p21(Cdkn1a). In contrast, short hairpin RNA-mediated depletion of RECK induced irreversible growth arrest along with several features of the Arf, Trp53 and Cdkn1a-dependent cellular senescence. Within 2 days of RECK depletion, we observed a transient increase in protein kinase B (AKT) and extracellular signal-regulated kinase (ERK) phosphorylation associated with an upregulated expression of cyclin D1, p19(Arf), Trp53, p21(Cdkn1a) and Sprouty 2. On further cultivation, RAS, AKT and ERK activities were then downregulated to a level lower than control, indicating that RECK depletion leads to a negative feedback to RAS signaling and subsequent cellular senescence. In addition, we observed that epidermal growth factor receptor (EGFR) activity was transiently upregulated by RECK depletion in MEFs, and continuously downregulated by RECK overexpression in colon cancer cells. These findings indicate that RECK is a novel modulator of EGFR signaling.
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys9) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-EGFR Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-p19 ARF Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse