Merck
  • Periodontal Pathogens Modulate Lipid Flux via Fatty Acid Binding Protein 4.
Periodontal Pathogens Modulate Lipid Flux via Fatty Acid Binding Protein 4.
Journal of dental research (2019-10-19)
D J Kim, J H Rho, B H Woo, J Y Joo, J Y Lee, J M Song, J H Lee, H R Park
ABSTRACT
A strong correlation between chronic periodontitis and systemic diseases (e.g., cardiovascular disease, metabolic disorders) has been suggested for several decades. However, the evidence supporting this correlation is restricted primarily to epidemiologic studies, with only a few experimental outcomes confirming such a correlation and providing information about the underlying molecular mechanisms. To reveal a correlation between periodontitis and systemic diseases as well as a relevant molecular pathway, we investigated the effects of Porphyromonas gingivalis and Fusobacterium nucleatum, which play roles in chronic periodontitis progression, on Raw264.7 and THP-1 macrophages. Infection with P. gingivalis or F. nucleatum significantly induced the expression of fatty acid binding protein 4 (FABP4), one of the most important adipokines that play a role in the progression of systemic diseases such as atherosclerosis and type 2 diabetes. Periodontal pathogen-induced FABP4 expression in macrophages promoted lipid uptake by these cells, as demonstrated by the diminished lipid accumulation in cells treated with an FABP4 inhibitor, BMS309403, or with knockdown of FABP4 expression. This periodontal pathogen-induced FABP4 expression was dependent on the JNK pathway, and JNK inhibition reduced lipid uptake by reducing FABP4 expression. Serum levels of antibodies against P. gingivalis correlated with serum FABP4 levels in humans, whereas no association occurred between F. nucleatum antibody titers and FABP4 levels. To our knowledge, this report is the first to experimentally demonstrate that periodontal pathogens stimulate lipid uptake in macrophages by modulating FABP4 expression. These findings strongly support the hypothesis that periodontitis may affect the progression of various systemic diseases.
Sigma-Aldrich
MISSION® esiRNA, targeting human FABP4