Merck
  • Bioaccessibility of micronutrients in fresh and frozen strawberry fruits grown under elevated carbon dioxide and temperature.
Bioaccessibility of micronutrients in fresh and frozen strawberry fruits grown under elevated carbon dioxide and temperature.
Food chemistry (2019-11-11)
Himali Balasooriya, Kithsiri Dasanayake, Said Ajlouni
ABSTRACT
Strawberry cultivar "San Andreas" was grown under ambient (400 ppm CO2, 25 °C) and elevated (950 ppm CO2, 30 °C) growth conditions. The strawberries were subjected to in vitro gastrointestinal digestion and colonic fermentation to examine the accessibility of polyphenols, vitamin C and folates in fresh and frozen fruits using HPLC-UV analyses. Results revealed that elevated CO2 and higher temperature enhanced the amounts of accessible bioactive compounds in strawberries. Bioaccessibility of pelargonidin-3-glucoside increased from 67% to 88% in strawberries grown under elevated growth. Fresh strawberries grown under ambient growth contained 93.09 ± 6.2 µg/100 g folates and 18.55 ± 0.5 mg/100 g vitamin C as bioaccessible fractions under fed state while, elevated growth enhanced soluble folates and vitamin C up to 188.63 ± 7.5 µg/100 g and 30.48 ± 0.3 mg/100 g, respectively. Fresh strawberries contained higher amounts of accessible micronutrients than frozen strawberries, while increased bile contents in intestinal fluid (fed state) facilitated the release of bioactive compounds to gastrointestinal fluid.
Sigma-Aldrich
5-Methyltetrahydrofolic acid disodium salt, ≥88% (UV-vis)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Supelco
Folinic acid calcium salt hydrate, analytical standard