Merck
  • Helix-Sense-Selective Polymerization of 3,5-bis(hydroxymethyl)phenylacetylene Rigidly Bearing Galvinoxyl Residues and Their Chiroptical Properties.
Helix-Sense-Selective Polymerization of 3,5-bis(hydroxymethyl)phenylacetylene Rigidly Bearing Galvinoxyl Residues and Their Chiroptical Properties.
Polymers (2019-11-27)
Zhichun Shi, Jianjun Wang, Masahiro Teraguchi, Toshiki Aoki, Takashi Kaneko
ABSTRACT
Four kinds of newly synthesized achiral phenylacetylenes bearing a phenylhydrogalvinoxyl residue at 4-position were polymerized by using a chiral rhodium catalyst system, [Rh(nbd)B(C6H5)4] or [Rh(nbd)Cl]2 catalysts in the presence of chiral (R)-(+)- or (S)-(-)-1-phenylethylamine ((R)- or (S)-PEA) cocatalysts. Poly(m-HGDHPA) and poly(m-HGTHPA) in THF showed Cotton signals at the absorption regions of the main chain and hydrogalvinoxyl in the circular dichroism (CD) spectra. It indicated that excess of one-handed helical polyacetylene backbone was induced by helix-sense-selective polymerization (HSSP) under the asymmetric conditions despite the achiral monomer, and the hydrogalvinoxyl moieties were also arranged to form one-handed helical structure. However, there was no Cotton effect for poly(p-HGDHPA) and poly(p-HGTHPA) because the intramolecular hydrogen bonding did not act well to stabilize the helical conformation. The hydrogalvinoxyl units of poly(m-HGDHPA) and poly(m-HGTHPA) were converted to the corresponding galvinoxyl radicals after treatment with PbO2. In the CD spectra of the polyradicals, the Cotton effects decreased depending on their static stability of helical conformation, suggesting that reversal conformation of the polymer chain arose.
Sigma-Aldrich
Ethynyltrimethylsilane, 98%
Sigma-Aldrich
4-Methoxycarbonylphenylboronic acid, ≥95%
Sigma-Aldrich
3-Methoxycarbonylphenylboronic acid