Merck
Porphyrin-phospholipid liposomes permeabilized by near-infrared light.
Nature communications (2014-04-05)
Kevin A Carter, Shuai Shao, Matthew I Hoopes, Dandan Luo, Bilal Ahsan, Vladimir M Grigoryants, Wentao Song, Haoyuan Huang, Guojian Zhang, Ravindra K Pandey, Jumin Geng, Blaine A Pfeifer, Charles P Scholes, Joaquin Ortega, Mikko Karttunen, Jonathan F Lovell
ABSTRACT
The delivery of therapeutic compounds to target tissues is a central challenge in treating disease. Externally controlled drug release systems hold potential to selectively enhance localized delivery. Here we describe liposomes doped with porphyrin-phospholipid that are permeabilized directly by near-infrared light. Molecular dynamics simulations identified a novel light-absorbing monomer esterified from clinically approved components predicted and experimentally demonstrated to give rise to a more stable porphyrin bilayer. Light-induced membrane permeabilization is enabled with liposomal inclusion of 10 molar % porphyrin-phospholipid and occurs in the absence of bulk or nanoscale heating. Liposomes reseal following laser exposure and permeability is modulated by varying porphyrin-phospholipid doping, irradiation intensity or irradiation duration. Porphyrin-phospholipid liposomes demonstrate spatial control of release of entrapped gentamicin and temporal control of release of entrapped fluorophores following intratumoral injection. Following systemic administration, laser irradiation enhances deposition of actively loaded doxorubicin in mouse xenografts, enabling an effective single-treatment antitumour therapy.
Product Description
Avanti
Cholesterol (ovine), Avanti Research - A Croda Brand
Avanti
16:0 Lyso PC, 1-palmitoyl-2-hydroxy-sn-glycero-3-phosphocholine, chloroform
Avanti
16:0 Lyso PC, Avanti Research - A Croda Brand
Avanti
18:0 PC (DSPC), Avanti Research - A Croda Brand
Avanti
18:0 PC (DSPC), Avanti Research - A Croda Brand
Avanti
18:0 PEG2000 PE, Avanti Research - A Croda Brand
Avanti
Hydro Soy PC, Avanti Research - A Croda Brand
Avanti
18:0 PG, Avanti Research - A Croda Brand
Avanti
18:0 PEG2000 PE, Avanti Research - A Croda Brand 880120C