Merck
  • Liposomes and drug-in-cyclodextrin-in-liposomes formulations encapsulating 17β-estradiol: An innovative drug delivery system that prevents the activation of the membrane-initiated steroid signaling (MISS) of estrogen receptor α.
Liposomes and drug-in-cyclodextrin-in-liposomes formulations encapsulating 17β-estradiol: An innovative drug delivery system that prevents the activation of the membrane-initiated steroid signaling (MISS) of estrogen receptor α.
International journal of pharmaceutics (2019-11-26)
Anne Gallez, Claudio Palazzo, Silvia Blacher, Ekaterine Tskitishvili, Agnès Noël, Jean-Michel Foidart, Brigitte Evrard, Christel Pequeux, Geraldine Piel
ABSTRACT
The encapsulation into liposomes of several types of molecules presents the advantages to protect the activity of these molecules and to target specific tissues. Nevertheless, a major obstacle remains the incomplete understanding of nano-bio interactions. Specifically, the impact that inclusion of drug into liposomes or of drug-in-cyclodextrin-in liposomes (DCL) could have on the molecular and cellular mechanism of drug action is largely unknown. As a proof of concept, we evaluated the impact of 17β-estradiol (E2) included into liposomes or DCL on estrogen receptor (ER)α signaling pathways. Indeed, ERα relays the pleiotropic actions of E2 in physiology and pathophysiology through two major pathways: (1) the genomic/nuclear effects associated to the transcriptional activity of the ERα and (2) the rapid/nongenomic/membrane-initiated steroid signaling (MISS) effects related to the induction of fast signaling pathways occurring when ERα is anchored to the plasma membrane. We evidenced that the inclusion of E2 into liposomes (Lipo-E2) or into DCL (DCL-E2) prevented the activation of the rapid/nongenomic/extranuclear/MISS pathway of ERα, while the activation of the genomic/nuclear pathway was maintained. These results support that Lipo-E2 and DCL-E2 could be a useful tool to delineate the complex molecular mechanisms associated to ERα. In conclusion, this study supports the notion that inclusion of drugs into liposomes or DCL could modify some specific pathways of their molecular and cellular mechanisms of action. These results emphasized that attention should be paid to nano-bio interactions induced by the use of nanovectors in medicine.
Product Description
Avanti
18:0 DDAB, Avanti Research - A Croda Brand 890810C
Avanti
18:0 DDAB, Avanti Research - A Croda Brand
Avanti
18:0 PEG2000 PE, Avanti Research - A Croda Brand
Avanti
18:0 PEG2000 PE, Avanti Research - A Croda Brand 880120C
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand
Sigma-Aldrich
1,2-Distearoyl-sn-glycero-3-phosphoethanolamine, ≥99%
Sigma-Aldrich
1,2-Dimyristoyl-d54-sn-glycero-3-phosphocholine, 98 atom % D, 97% (CP)