Merck
  • Shc regulates epidermal growth factor-induced activation of the JNK signaling pathway.
Shc regulates epidermal growth factor-induced activation of the JNK signaling pathway.
The Journal of biological chemistry (1999-07-10)
A Hashimoto, M Kurosaki, N Gotoh, M Shibuya, T Kurosaki
ABSTRACT
Two adaptor molecules, Grb2 and Shc, have been implicated in the extracellular signal-regulated kinase (ERK) activation by receptor tyrosine kinases such as the epidermal growth factor receptor (EGFR). Here we show that the EGF-mediated ERK activation is abolished by loss of Grb2, whereas this response is not affected by loss of Shc. Conversely, the EGF-mediated c-Jun N-terminal kinase (JNK) activation is dependent on Shc, but not Grb2. These findings strongly support distinct roles for Grb2 and Shc in controlling ERK and JNK activation after EGF stimulation.