Merck
  • Rab46 integrates Ca2+ and histamine signaling to regulate selective cargo release from Weibel-Palade bodies.
Rab46 integrates Ca2+ and histamine signaling to regulate selective cargo release from Weibel-Palade bodies.
The Journal of cell biology (2019-05-17)
Katarina T Miteva, Lucia Pedicini, Lesley A Wilson, Izzy Jayasinghe, Raphael G Slip, Katarzyna Marszalek, Hannah J Gaunt, Fiona Bartoli, Shruthi Deivasigamani, Diego Sobradillo, David J Beech, Lynn McKeown
ABSTRACT
Endothelial cells selectively release cargo stored in Weibel-Palade bodies (WPBs) to regulate vascular function, but the underlying mechanisms are poorly understood. Here we show that histamine evokes the release of the proinflammatory ligand, P-selectin, while diverting WPBs carrying non-inflammatory cargo away from the plasma membrane to the microtubule organizing center. This differential trafficking is dependent on Rab46 (CRACR2A), a newly identified Ca2+-sensing GTPase, which localizes to a subset of P-selectin-negative WPBs. After acute stimulation of the H1 receptor, GTP-bound Rab46 evokes dynein-dependent retrograde transport of a subset of WPBs along microtubules. Upon continued histamine stimulation, Rab46 senses localized elevations of intracellular calcium and evokes dispersal of microtubule organizing center-clustered WPBs. These data demonstrate for the first time that a Rab GTPase, Rab46, integrates G protein and Ca2+ signals to couple on-demand histamine signals to selective WPB trafficking.
Sigma-Aldrich
(±)-Epinephrine hydrochloride
Sigma-Aldrich
Nocodazole Ready Made Solution, 5 mg/mL, DMSO solution
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine (TMB) Liquid Substrate System for ELISA, peroxidase substrate