Merck
  • Recombinant Mycobacterium leprae protein associated with entry into mammalian cells of respiratory and skin components.
Recombinant Mycobacterium leprae protein associated with entry into mammalian cells of respiratory and skin components.
Journal of dermatological science (2007-02-24)
Naoya Sato, Takao Fujimura, Mikio Masuzawa, Yasuko Yogi, Masanori Matsuoka, Maho Kanoh, Lee W Riley, Kensei Katsuoka
ABSTRACT
The transmission of Mycobacterium leprae, the causative pathogen of leprosy, has been postulated to occur mainly through upper respiratory route rather than skin-to-skin contact via minor injuries. The M. leprae genome contains mce1A gene, which encodes a putative mammalian cell entry protein. However, to date, there have been no functional analyses of the M. leprae mce1A gene product. The aim of this study was to elucidate a possible relationship between this transmission mechanism and the mce1A gene product. We analyzed the cell uptake activity in vitro of polystyrene latex beads coated with a purified recombinant (r-) protein expressed by a 849-bp locus within the mce1A gene. The r-protein promoted uptake of the beads into human nasal epithelial cells derived from nasal polyps, human bronchial epithelial cell line, normal human dermal fibroblasts, normal human microvascular endothelial cells and normal human keratinocytes cultured at 0.01 mM extracellular calcium concentration [Ca]; no uptake occurred with keratinocytes cultured at 1.2mM [Ca]. These results suggest that the mce1A gene product can mediate M. leprae entry into respiratory epithelial cells as their natural target cells, which may be the main mode of transmission. Endothelial cells, on the other hand, may serve as the reservoir of the bacilli for long-term infection. The M. leprae Mce1A protein has potential important implications for mode of transmission and pathogenesis of leprosy.
Sigma-Aldrich
Latex beads, amine-modified polystyrene, fluorescent blue, aqueous suspension, 0.05 μm mean particle size
Sigma-Aldrich
Latex beads, amine-modified polystyrene, fluorescent yellow-green, aqueous suspension, 1.0 μm mean particle size
Sigma-Aldrich
Latex beads, amine-modified polystyrene, fluorescent blue, aqueous suspension, 2.0 μm mean particle size