Merck
  • Novel antitumor agents CI-920, PD 113,270 and PD 113,271. I. Taxonomy, fermentation and biological properties.
Novel antitumor agents CI-920, PD 113,270 and PD 113,271. I. Taxonomy, fermentation and biological properties.
The Journal of antibiotics (1983-12-01)
J B Tunac, B D Graham, W E Dobson
ABSTRACT
CI-920 (PD 110,161) and two analogues (PD 113,270 and PD 113,271) are novel antitumor compounds produced by a new actinomycete characterized as Streptomyces pulveraceus subsp. fostreus ATCC 31906. The antitumor compounds are predominantly produced during the stationary (idiophase) growth phase of the organism. CI-920 is active versus the murine P388 lymphocytic and L1210 lymphoid leukemia with T/C values of 246 and 207, respectively. This compound has no significant antimicrobial activity.
Sigma-Aldrich
Fostriecin sodium salt from Streptomyces pulveraceus, ≥98% (HPLC)