- A molecular mechanism of integrin crosstalk: alphavbeta3 suppression of calcium/calmodulin-dependent protein kinase II regulates alpha5beta1 function.
A molecular mechanism of integrin crosstalk: alphavbeta3 suppression of calcium/calmodulin-dependent protein kinase II regulates alpha5beta1 function.
Many cells express more than one integrin receptor for extracellular matrix, and in vivo these receptors may be simultaneously engaged. Ligation of one integrin may influence the behavior of others on the cell, a phenomenon we have called integrin crosstalk. Ligation of the integrin alphavbeta3 inhibits both phagocytosis and migration mediated by alpha5beta1 on the same cell, and the beta3 cytoplasmic tail is necessary and sufficient for this regulation of alpha5beta1. Ligation of alpha5beta1 activates the calcium- and calmodulin-dependent protein kinase II (CamKII). This activation is required for alpha5beta1-mediated phagocytosis and migration. Simultaneous ligation of alphavbeta3 or expression of a chimeric molecule with a free beta3 cytoplasmic tail prevents alpha5beta1-mediated activation of CamKII. Expression of a constitutively active CamKII restores alpha5beta1 functions blocked by alphavbeta3-initiated integrin crosstalk. Thus, alphavbeta3 inhibition of alpha5beta1 activation of CamKII is required for its role in integrin crosstalk. Structure-function analysis of the beta3 cytoplasmic tail demonstrates a requirement for Ser752 in beta3-mediated suppression of CamKII activation, while crosstalk is independent of Tyr747 and Tyr759, implicating Ser752, but not beta3 tyrosine phosphorylation in initiation of the alphavbeta3 signal for integrin crosstalk.