Merck
  • Antimetastatic effect of the pharmacological inhibition of serine/arginine-rich protein kinases (SRPK) in murine melanoma.
Antimetastatic effect of the pharmacological inhibition of serine/arginine-rich protein kinases (SRPK) in murine melanoma.
Toxicology and applied pharmacology (2018-08-24)
Gabriela Alves Moreira, Graziela Domingues de Almeida Lima, Raoni Pais Siqueira, Marcus Vinícius de Andrade Barros, Abraham Landry Mahuvi Adjanohoun, Viviane Corrêa Santos, Éverton de Almeida Alves Barbosa, Robson Kriiger Loterio, Janine Cerqueira de Paiva, Victor Hugo Sousa Gonçalves, Lívia Cristina de Souza Viol, Eduardo de Almeida Marques-da-Silva, Abelardo Silva Júnior, Márcia Rogéria Almeida, Juliana Lopes Rangel Fietto, Mariana Machado-Neves, Rafaela Salgado Ferreira, Róbson Ricardo Teixeira, Gustavo Costa Bressan
ABSTRACT
The Serine/arginine-rich protein kinases (SRPK) are involved in pre-mRNA splicing control through the phosphorylation of the SR protein family of splicing factors. Over the last years, several studies have shown the relevance of SRPK for human cancers and their potential as promising drug targets. In this context, we have previously selected three trifluoromethyl arylamides (named here as SRVIC24, SRVIC30 and SRVIC36) with improved in vitro antileukemia effect and ability of impairing the cellular activity of SRPK. Given the increasing amount of reports on the implication of these kinases in metastatic cancers, in this study, we have evaluated the antimetastatic effect of these compounds and the known SRPK inhibitor (SRPIN340) on a murine model of metastatic melanoma. The compounds were able to impact the melanoma cell metastatic behavior by decreasing migration, invasion, adhesion, and colony formation in in vitro assays. Also, they presented antimetastatic in vivo activity, without apparent signs of systemic toxicity after treatments, as revealed by the histology of organs and analysis of key serum biochemical markers. Moreover, the effect of the treatments on SRPK1 nuclear translocation and SR protein phosphorylation was observed. Finally, molecular docking studies were carried out to gain structural information on the SRPK-compound complexes. Together, these data suggest that SRPK pharmacological inhibition should be considered as an interesting therapeutic strategy against metastatic cancers.
Sigma-Aldrich
SRPIN340, ≥98% (HPLC)
Sigma-Aldrich
Toluidine Blue, 8.74% (ZN (THEORY))