Skip to Content
Merck
Veratraldehyde ≥98%, FG

Veratraldehyde

Veratraldehyde ≥98%, FG
Synonym(s):
3,4-Dimethoxybenzaldehyde, Methylvanillin, NSC 24521, NSC 8500, Vanillin methyl ether, Veratraldehyde
Linear Formula:
(CH3O)2C6H3CHO
CAS Number:
Molecular Weight:
166.17
EC Number:

Products

Product Category
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing