
Trospium impurity C

Synonym(s):
(1R,3r,5S)-3-Hydroxyspiro[8-azoniabicyclo[3.2.1]octane-8,1′-pyrrolidinium] chloride
Empirical Formula (Hill Notation):
C11H20ClNO
CAS Number:
Molecular Weight:
217.74
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing