
Triamcinolone acetonide Related Compound B

Synonym(s):
(11β,16α)-9-Fluoro-11,21-dihydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregna-1,4,14-triene-3,20-dione
Empirical Formula (Hill Notation):
C24H29FO6
CAS Number:
Molecular Weight:
432.48
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing