Skip to Content
Merck
trans-Styrylacetic acid 96%

trans-Styrylacetic acid

trans-Styrylacetic acid 96%
Synonym(s):
4-Phenyl-3-butenoic acid
Linear Formula:
C6H5CH=CHCH2CO2H
CAS Number:
Molecular Weight:
162.19

Products

Product Category
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing