Skip to Content
Merck
Poly(3,4-ethylenedioxythiophene), tetramethacrylate end-capped solution 0.5 wt. % (dispersion in nitromethane), contains p-toluenesulfonate as dopant

PEDOT

Poly(3,4-ethylenedioxythiophene), tetramethacrylate end-capped solution 0.5 wt. % (dispersion in nitromethane), contains p-toluenesulfonate as dopant
Synonym(s):
PEDOT, Poly(2,3-dihydrothieno-1,4-dioxin), C12-PEG-block-PEDOT-block-PEG-C12, PEDOT:PEG, PEDOT, tetramethacrylate end-capped
Linear Formula:
(C6H4O2S)n ·xH2O
CAS Number: