Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nickel oxide
Linear Formula:
NiO
CAS Number:
1313-99-1
Molecular Weight:
74.69
EC Number:
215-215-7
Products
Product Category
anode materials (2)
oxides (2)
Brand
Sigma-Aldrich (2)
Sustainability
Color
Feature
Formula Weight
Physical Form
Purity
Chemical Composition
Quality Segment
Application
Particle Size (µm)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
203882
Nickel oxide,
Ni(II), 99.99% trace metals basis
Expand
View Pricing
637130
Nickel oxide,
Ni(II), nanopowder, <50 nm particle size (TEM), 99.8% trace metals basis
Expand
View Pricing
399523
Nickel oxide,
Ni(II), green, −325 mesh, 99%
Expand
View Pricing
481793
Nickel oxide,
Ni(II), ≥99.995% trace metals basis
Expand
View Pricing