Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nalgene
®
HDPE Biotainer
®
Bottles
Products
Product Category
bottles (3)
Manufacturer
Capacity (mL)
Color
Package Size
Sterility
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
TMO3410-42
Nalgene
®
HDPE Biotainer
®
Bottles,
clear polycarbonate, 10000 mL capacity, case of 2 ea
Expand
View Pricing
TMO3405-42
Nalgene
®
HDPE Biotainer
®
Bottles,
clear polycarbonate, 5000 mL capacity, case of 6 ea bulk pack
Expand
View Pricing
TMO3415-42
Nalgene
®
HDPE Biotainer
®
Bottles,
clear PETG, 5000 mL capacity, case of 6 ea bulk pack
Expand
View Pricing