Skip to Content
Merck
vibrant-m

Montelukast racemate

vibrant-m
Synonym(s):
Montelukast dicyclohexylamine racemate
Empirical Formula (Hill Notation):
C35H36ClNO3S · C12H23N
Molecular Weight:
767.50

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing