
Monoethylglycinexylidine

Synonym(s):
MEGX, MGEX, Monoethylglycinexylidide, N-(2,6-dimethylphenyl)-2-(ethylamino)acetamide, Norlidocaine
Empirical Formula (Hill Notation):
C12H18N2O
CAS Number:
Molecular Weight:
206.28
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing