
Meat Peptone for Biotechnology and Microbiology

Synonym(s):
Peptone from meat, Peptone from bovine meat
CAS Number:
Products
Product Category
Brand
Biological Source
Color
pH Value
Physical Form
Sterilization Compatibility
Tissue
Chemical Composition
Quality Segment
Application
Microorganism Suitability
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing