Skip to Content
Merck
Lactose ureide-15N2 ≥98 atom % 15N, ≥98% (CP)

Lactose ureide-15N2

Lactose ureide-15N2 ≥98 atom % 15N, ≥98% (CP)
Empirical Formula (Hill Notation):
C13H2415N2O11
Molecular Weight:
386.32

Products

Product Category
Formula Weight
Physical Form
Purity
Isotope
Showing 1-1 of 1
Compare
Product Number
Description
Pricing