
Hydroflumethiazide

Synonym(s):
3,4-Dihydro-6-(trifluoromethyl)-2H-1,2,4-benzothiadiazine-7-sulfonamide 1,1-dioxide
Empirical Formula (Hill Notation):
C8H8F3N3O4S2
CAS Number:
Molecular Weight:
331.29
Products
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing