
Giemsa Stain

Synonym(s):
Azure eosin methylene blue, Giemsa solution
CAS Number:
EC Number:
Products
Product Category
Color
Physical Form
Quality Segment
Application
Microorganism Suitability
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing