Skip to Content
Merck
Fluid A - Ready-to-use Rinse Fluid bottle capacity 1000 mL, bottle filling volume 800 mL, closure type, Red screw cap with 3 septa, pack of 6 bottles

Fluid A - Ready-to-use Rinse Fluid

Fluid A - Ready-to-use Rinse Fluid bottle capacity 1000 mL, bottle filling volume 800 mL, closure type, Red screw cap with 3 septa, pack of 6 bottles

Products

Product Category
Brand
Physical Form
Sterilization Compatibility
Agency
Chemical Composition
Capacity (mL)
Compatibility
Quality Segment
Closure Type
Application
Package Size
Sterility
Sterilization Method
Technique
Showing 1-15 of 15
Compare
Product Number
Description
Pricing