
Febantel for system suitability

Synonym(s):
Febantel, N-{2-[2,3-Bis-(methoxycarbonyl)-guanido]-5-(phenylthio)-phenyl}-2-methoxyacetamide
Empirical Formula (Hill Notation):
C20H22N4O6S
CAS Number:
Molecular Weight:
446.48
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing