Skip to Content
Merck
DOTAP chloride ≥98% (TLC), powder

DOTAP chloride

DOTAP chloride ≥98% (TLC), powder
Synonym(s):
N-[1-(2,3-Dioleoyloxy)propyl]-N,N,N-trimethylammonium chloride
Empirical Formula (Hill Notation):
C42H80ClNO4
CAS Number:
Molecular Weight:
698.54

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing