
Clear Thioglycollate Medium

Products
Product Category
Brand
Physical Form
Sterilization Compatibility
Agency
Capacity (mL)
Compatibility
Quality Segment
Closure Type
Application
Microorganism Suitability
Package Size
Sterility
Sterilization Method
Showing 1-5 of 5
Compare
Product Number
Description
Pricing