Skip to Content
Merck
Tryptone Plus for biotechnological purposes

Casein Peptone suitable for biotechnology and microbiology

Tryptone Plus for biotechnological purposes
Synonym(s):
Peptone from casein (Tryptone), Peptone from casein, Peptone from casein pancreatically digested
CAS Number:
EC Number:

Products

Product Category
Brand
Biological Source
Color
pH Value
Physical Form
Tissue
Chemical Composition
Quality Segment
Application
Microorganism Suitability
Package Size
Special Grade
Showing 1-16 of 16
Compare
Product Number
Description
Pricing